Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methylimidazole-2-sulphonyl chloride
1-Methylimidazole-2-sulphonyl chloride, Sigma-Aldrich®
1-Methylimidazole-2-sulphonyl chloride, Sigma-Aldrich®
SIAL718211-1G
:
Merck
:
1-Methylimidazole-2-sulphonyl chloride, Sigma-Aldrich®
SIAL718211-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
55694-81-0
:
C₄H₅ClN₂O₂S
:
MFCD16621450