Methyl diphenylphosphinite, Sigma-Aldrich®
Catalog # SIAL149497-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.22 g/mol
:M-Clarity™ quality level = MQ100
EINECS:223-683-9
CAS Number:4020-99-9
Formula:C₁₃H₁₃OP
MDL Number:MFCD00048025