

(3-Aminopropyl)dimethylmethoxysilane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31024-26-7
:
Formula:C₆H₁₇NOSi
MDL Number:MFCD01861689
Boiling Pt:87 °C (50 mmHg)
Flash Pt:38 °C
UN:2924
ADR:3,III
:GHS02
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4300 to 1.4320 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together