
(S)-(-)-1-Boc-3-methylpiperazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:147081-29-6
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD02683204
:200.28 g/mol
Melting Pt:38…41 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together