Bathocuproin sulphonate disodium salt hydrate 97%
:52698-84-7
:
:C₂₆H₁₈N₂Na₂O₆S₂·H₂O
:MFCD00149974
:300 °C
:564.55 g/mol (anhy)
:Ambient
:258-111-7
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤9 %
- HPLC:≥96.0 % (expressed as the sum of ortho-ortho, meta-para and para-para-substituted bathocuproine disulfonate)
Frequently Bought Together