2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, Sigma-Aldrich®
Catalog # SIALN5504-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:246.738 g/mol
:M-Clarity™ quality level = MQ100
EINECS:208-989-2
CAS Number:550-99-2
Formula:C₁₄H₁₅ClN₂
MDL Number:MFCD00012554