Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Cyanophenyl)-2,5-dimethylpyrrole ≥98%
Irritant
1-(4-Cyanophenyl)-2,5-dimethylpyrrole ≥98%
Catalog # H61178.06
CAS Number:  
Irritant
1-(4-Cyanophenyl)-2,5-dimethylpyrrole ≥98%
Catalog # H61178.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :196.25 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:119516-86-8
Formula:C₁₃H₁₂N₂
MDL Number:MFCD03228548