Order Entry
Czech Republic
ContactUsLinkComponent
Choline hydroxide 45% in methanol, Sigma-Aldrich®
Choline hydroxide 45% in methanol, Sigma-Aldrich®
Catalog # SIAL333018-1KG
Supplier:  Merck
CAS Number:  
Choline hydroxide 45% in methanol, Sigma-Aldrich®
Catalog # SIAL333018-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :121.18 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:204-625-1
CAS Number:123-41-1
Formula:C₅H₁₅NO₂
MDL Number:MFCD00002831