
2,5-Diaminohydroquinone dihydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24171-03-7
:
Formula:C₆H₁₀Cl₂N₂O₂
MDL Number:MFCD00239416
:213.06 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together