
1-Methylimidazole-5-carbaldehyde ≥97%
Catalog # H27338.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:39021-62-0
:
Formula:C₅H₆N₂O
MDL Number:MFCD00030439
:110.12 g/mol
Melting Pt:52…57 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient