Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium bromide
Aluminium bromide 1 mol/l in dibromomethane, Sigma-Aldrich®
Aluminium bromide 1 mol/l in dibromomethane, Sigma-Aldrich®
SIAL250066-100ML
:
Merck
:
Aluminium bromide 1 mol/l in dibromomethane, Sigma-Aldrich®
SIAL250066-100ML
:
Merck
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
266.69 g/mol
:
M-Clarity™ quality level = MQ100
:
231-779-7
:
7727-15-3
:
AlBr₃
:
MFCD00003421