
(2-Bromo-5-fluorophenyl)acetic acid 96%
Catalog # H62005.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:739336-26-6
:
Formula:C₈H₆BrFO₂
MDL Number:MFCD03094440
Melting Pt:113…114 °C
:233.03 g/mol
:GHS07
Storage Temperature:Ambient