Methyl-2-methyl-3-nitrobenzoate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59382-59-1
:
Formula:C₉H₉NO₄
MDL Number:MFCD00082564
:195.17 g/mol
Melting Pt:62…65 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together