
2-Bromo-3,4-difluoroaniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1092349-87-5
:
Formula:C₆H₄BrF₂N
MDL Number:MFCD11035924
:208.00 g/mol
ADR:6.1,III
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together