Order Entry
Czech Republic
ContactUsLinkComponent
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
Catalog # ABCRAB204671
Supplier:  abcr
CAS Number:  
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
Catalog # ABCRAB204671
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mL

Specifications

About this item

 :
 :468.2800 g/mol
EINECS:288-657-1
CAS Number:85857-16-5
Formula:C₁₁H₁₃F₁₃O₃Si
MDL Number:MFCD00153261