
3-Iodo-1H-pyrazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4522-35-4
:
Formula:C₃H₃IN₂
MDL Number:MFCD12022324
:193.98 g/mol
Melting Pt:69…72 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together