Order Entry
Czech Republic
ContactUsLinkComponent
3-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester 96%
3-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester 96%
Catalog # H62811.03
CAS Number:  
3-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester 96%
Catalog # H62811.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:445303-67-3
 :
Formula:C₁₃H₁₅BF₄O₂
MDL Number:MFCD09878544
 :290.06 g/mol
Storage Temperature:Ambient