2-Cyano-4-(trifluoromethyl)phenylboronic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1218790-84-1
:
Formula:C₈H₅BF₃NO₂
MDL Number:MFCD13195679
:214.94 g/mol
Storage Temperature:Ambient
Frequently Bought Together