Order Entry
Czech Republic
ContactUsLinkComponent
 
Bromothymol blue ACS
Catalog # 0431-25G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromothymol blue ACS
Catalog # 0431-25G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Clarity of solution:
    Pass
  • Visual transition interval (pH 6.0 − 7.6):
    6.0(yellow) − 7.6(blue)

Specification Test Results

About this item

 :
Merck Index:13,01430
Melting Pt:204 − 202 °C
 :624.39 g/mol
Melting Pt:204…202 °C
Storage Temperature:Ambient
EINECS:200-971-2
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872