ContactUsLinkComponent
Stoppers for crimp caps, N 20
Stoppers for crimp caps, N 20
Catalog # MANA702931.1
Supplier:  MACHEREY-NAGEL
undefined
Stoppers for crimp caps, N 20
Catalog # MANA702931.1
Supplier:  MACHEREY-NAGEL
Specifications
  • Type:
    Bromobutyl red, 45° shore A hardness
  • Seq:
    44
  • Pk:
    100