ContactUsLinkComponent
Racks for Leighton tissue culture tubes
Racks for Leighton tissue culture tubes
Catalog # BELC1917-00016
Supplier:  BELLCO GLASS
undefined
Racks for Leighton tissue culture tubes
Catalog # BELC1917-00016
Supplier:  BELLCO GLASS
Specifications
  • For tubes:
    125 - 150 mm
  • No. of Holes:
    36
  • Description:
    Self locking, for Leighton type tissue culture tubes
  • Pk:
    1
Specifications
About this item
Stainless steel.
  • Spring clip prevents tube from moving in rack
  • Excellent chemical resistance
  • Excellent temperature resistance
For Leighton type culture tubes.