VWR® LB, Broth, Liquid, for Biotechnology
Catalog # J833-1L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Description:LB broth, liquid
- Environmentally Preferable:
Specifications
About this item
LB Broth, liquid is used for the propagation and maintenance of E. coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
- Ready to use liquid media
- Clear, yellow to brown liquid
One L of media contains 10 g sodium chloride, 10 g tryptone/case in peptone, 5 g yeast extract, bacteriological.
No growth detected in bioburden (Milliflex) (EQC) test.
pH (25 °C): 6,65 to 7,15.