Order Entry
Czech Republic
ContactUsLinkComponent
Periodic acid solution, Q Path® for microscopy
Periodic acid solution, Q Path® for microscopy
Catalog # 10047024.
Supplier:  VWR Chemicals
CAS Number:  
undefined
Periodic acid solution, Q Path® for microscopy
Catalog # 10047024.
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Pouch
  • Pk:
    450 mL
  • Seq:
    2675

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Density (20/4):
    0,99 - 1,01

Specification Test Results

About this item

 :
 :227.94 g/mol
Storage Temperature:Ambient
EINECS:233-937-0
 :1
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344