Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Methyl purple 1 g/l in aqueous solution indicator
  HACH2193432
 :  Hach
 :  
 
Methyl purple 1 g/l in aqueous solution indicator
  HACH2193432
 :  Hach
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

 :
 :1391.41 g/mol
 :207-776-1
 :GHS09
 :1340-02-9
 :C₆₇H₆₂N₈Na₄O₁₄S₃
 :MFCD00070843