Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dichloro-4-methylpyridine
2,6-Dichloro-4-methylpyridine
Catalog # APOSOR60099-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-4-methylpyridine
Catalog # APOSOR60099-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.02 g/mol
CAS Number:39621-00-6
Formula:C₆H₅Cl₂N
MDL Number:MFCD09264302