Order Entry
Czech Republic
ContactUsLinkComponent
5-Isopropyl-1,3-cyclohexanedione ≥99% (by GC), Sigma-Aldrich®
5-Isopropyl-1,3-cyclohexanedione ≥99% (by GC), Sigma-Aldrich®
Catalog # SIAL343447-5G
Supplier:  Merck
CAS Number:  
5-Isopropyl-1,3-cyclohexanedione ≥99% (by GC), Sigma-Aldrich®
Catalog # SIAL343447-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :154.21 g/mol (anhy)
 :M-Clarity™ quality level = MQ100
CAS Number:18456-87-6
Formula:C₉H₁₄O₂·H₂O
MDL Number:MFCD00075263