Order Entry
Czech Republic
ContactUsLinkComponent
2-Amino-5-chloro-6-methylpyridine 97%
Corrosive
Irritant
2-Amino-5-chloro-6-methylpyridine 97%
Catalog # H58501.06
CAS Number:  
Corrosive
Irritant
2-Amino-5-chloro-6-methylpyridine 97%
Catalog # H58501.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :142.59 g/mol
Melting Pt:70…75 °C
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:36936-23-9
Formula:C₆H₇ClN₂
MDL Number:MFCD09453335