5-Phenyl-1,3-cyclohexanedione ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:493-72-1
:
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00051846
:188.23 g/mol
Melting Pt:∼ 185 °C (dec.)
Storage Temperature:Ambient
Related Information
Frequently Bought Together