Order Entry
Czech Republic
ContactUsLinkComponent
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
Catalog # SIAL292966-5G
Supplier:  Merck
CAS Number:  
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
Catalog # SIAL292966-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :348.29 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:252-425-8
CAS Number:35193-63-6
Formula:C₂₀H₁₃O₄P
MDL Number:MFCD00010045