2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-27-7
:
Formula:C₁₇H₂₉NO
MDL Number:MFCD00026283
:263.4183 g/mol
:M-Clarity™ quality level = MQ200
EINECS:201-816-1
Frequently Bought Together