(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol, Sigma-Aldrich®
Catalog # SIAL717746-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:638.7500 g/mol
CAS Number:361342-50-9
Formula:C₄₈H₃₀O₂
MDL Number:MFCD09027270