ContactUsLinkComponent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 0.1 mol/l in aqueous solution, Millipore®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 0.1 mol/l in aqueous solution, Millipore®
  SIAL19899-1ML-F
 :  Merck
 :  
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 0.1 mol/l in aqueous solution, Millipore®
  SIAL19899-1ML-F
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070
 :
 :614.89 g/mol
 :Refrigerator
 :75621-03-3
 :C₃₂H₅₈N₂O₇S
 :MFCD00081083