


Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16761-83-4
:
Formula:C₁₅H₃₅O₅Ta
MDL Number:MFCD00145347
:476.39 g/mol
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS02
:GHS08
:GHS07
Related Information
Frequently Bought Together