
Sodium hydroxide monohydrate ≥99.996% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12179-02-1
:
Formula:HNaO·1H₂O
MDL Number:MFCD00149229
:58.012 g/mol
Merck Index:14,8627
Boiling Pt:1390 °C
Melting Pt:318.4 °C
Density:2.13
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
:GHS05
Related Information
Frequently Bought Together