Order Entry
Czech Republic
ContactUsLinkComponent
 
Magnesium acetate tetrahydrate 98.0-102.0%, AnalaR NORMAPUR®
 
undefined
Magnesium acetate tetrahydrate 98.0-102.0%, AnalaR NORMAPUR®
 

 

  • Assay:
    98.0 - 102.0 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ba (Barium):
    Max. 10 ppm
  • Ca (Calcium):
    Max. 100 ppm
  • Fe (Iron):
    Max. 5 ppm
  • K (Potassium):
    Max. 50 ppm
  • Mn (Manganese):
    Max. 10 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Sr (Strontium):
    Max. 50 ppm

 

Frequently Bought Together