HPLC columns, Hypersil™ ODS-2 C18
Supplier: Thermo Fisher Scientific
Certificates
About this item
Hypersil™ ODS-2 C18 columns are well established and referenced in HPLC methods. Hypersil™ stationary phases are recognised as an industry standard in HPLC.
- Reproducibility and high selectivity over wide range of applications
- Dependable L1 and well tested
Frequently Bought Together