Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Sigma-Aldrich®
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL2105-50ML
Supplier:  Merck
CAS Number:  
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL2105-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,III
 :M-Clarity™ quality level = MQ200
EINECS:215-185-5
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548