Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethylene glycol
Ethylene glycol 5 mol/l in aqueous solution, Millipore®
Ethylene glycol 5 mol/l in aqueous solution, Millipore®
SIAL61941-1ML-F
:
Merck
:
Ethylene glycol 5 mol/l in aqueous solution, Millipore®
SIAL61941-1ML-F
:
Merck
:
Pack type:
Glass bottle
Pk:
1 mL
Seq:
2070
:
:
62.07 g/mol
:
Refrigerator
:
203-473-3
:
107-21-1
:
HOCH₂CH₂OH
:
MFCD00002885