Order Entry
Czech Republic
ContactUsLinkComponent
Sulphuric acid 15% (v/v) in aqueous solution
Corrosive
Sulphuric acid 15% (v/v) in aqueous solution
Catalog # 42554.K2
CAS Number:  
Corrosive
Sulphuric acid 15% (v/v) in aqueous solution
Catalog # 42554.K2
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589