Sodium thiosulphate 0.01 mol/l (0.01 N) in aqueous solution
Catalog # SCARSO07331000
Supplier: Scharlau
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499