
Bromocresol green 0.1% in ethanol
Supplier: MACH
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Storage Temperature:Ambient
EINECS:200-972-8
UN:1170
ADR:3,II
:GHS02
Frequently Bought Together