Order Entry
Czech Republic
ContactUsLinkComponent
Nitric acid 5 mol/l (5 N)
Korozi způsobující
Nitric acid 5 mol/l (5 N)
Catalog # CMPLZ14224218.5L
Číslo CAS:  
Korozi způsobující
Nitric acid 5 mol/l (5 N)
Catalog # CMPLZ14224218.5L
Číslo CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifikace

  • Pk:
    5 L
  • Seq:
    5190

Specifikace

O tomto produktu

 :
 :63.01 g/mol
Teplota pro skladování:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
Číslo CAS:7697-37-2
Vzorec:HNO₃
MDL číslo:MFCD00011349