Conductivity standard 1413 µS/cm (25 °C; 0,01 mol/L; KCl)
Catalog # SCARPA01010500
Supplier: Scharlau
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Reference temperature:25 °C
- Description:Conductivity standard 1413 µS/cm (0,01 mol/l; KCl)