
Methyl-4-amino-3-chlorobenzoate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84228-44-4
:
Formula:C₈H₈ClNO₂
MDL Number:MFCD01320686
:185.61 g/mol
Melting Pt:106…110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Beige to light brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:106 to 110 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together