Methyl-2-fluoroisonicotinate, Sigma-Aldrich®
Catalog # SIAL697869-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:155.1300 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:455-69-6
Formula:C₇H₆FNO₂
MDL Number:MFCD11656063