Methyl-4-bromo-3-methylbenzoate, Sigma-Aldrich®
Catalog # SIAL532878-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:229.07 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:148547-19-7
Formula:C₉H₉BrO₂
MDL Number:MFCD00673014