Ethyl trifluoropyruvate, Sigma-Aldrich®
Catalog # SIAL510254-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:170.09 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:13081-18-0
Formula:C₅H₅F₃O₃
MDL Number:MFCD00114935