Order Entry
Czech Republic
ContactUsLinkComponent
Hydrogen chloride 5->6 N in 2-propanol, pure
Irritant
Corrosive
Flammable
Hydrogen chloride 5->6 N in 2-propanol, pure
Certificates
undefined
Irritant
Corrosive
Flammable
Hydrogen chloride 5->6 N in 2-propanol, pure
Certificates

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Water:
    ≤1.0 %
  • Titration with NaOH:
    5 to 6 N
  • NMR (31P-NMR):
    Consistent with structure
  • Elemental Analysis:
    41.9 to 45.4 % C

Specification Test Results

Related Information

Frequently Bought Together