


Hydrogen chloride ∼0.5 M in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Density:0.8
Flash Pt:12 °C (54 °F)
EINECS:231-595-7
UN:1992
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together