


Chlorovodík 5->6 M in 2-propanol, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:0,909
Flash Pt:11 °C (52 °F)
Storage Temperature:Refrigerator
EINECS:231-595-7
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
Related Information
Frequently Bought Together